missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC6A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143620
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SLC6A4 Polyclonal Antibody for ICC/IF, IHC, Western Blot
SERT is an integral membrane protein that transports the neurotransmitter serotonin from synaptic spaces into presynaptic neurons. The encoded protein terminates the action of serotonin and recycles it in a sodium-dependent manner. This protein is a target of psychomotor stimulants, such as amphetamines and cocaine, and is a member of the sodium:neurotransmitter symporter family. A repeat length polymorphism in the promoter of this gene has been shown to affect the rate of serotonin uptake and may play a role in sudden infant death syndrome, aggressive behavior in Alzheimer disease patients, and depression-susceptibility in people experiencing emotional trauma.
Spezifikation
| SLC6A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC6A4 | |
| 5 OCD1; 5HT transporter; 5HTT; 5-HTT; 5-HTTLPR; 5-hydroxytryptamine (serotonin) transporter; AI323329; HGNC:11050; hSERT; HTT; Na+/Cl- dependent serotonin transporter; OCD1; OTTHUMP00000163633; serotonin transporter; serotonin transporter 1; Sert; SERT1; Slc6a4; Sodium-dependent serotonin transporter; Sodium-dependent serotonin transporter (5HT transporter) (5HTT); solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); Solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); solute carrier family 6 (neurotransmitter transporter), member 4; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; solute carrier family 6 member 4; solute carrier family 6, member 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15567, 25553, 282365, 6532 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P31645, P31652, Q60857, Q9XT49 | |
| SLC6A4 | |
| KSITPETPTEIPCGDIRLNAV coupled to KLH. | |
| 100 μL | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur