missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC6A8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15525272
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15525272 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15525272 Supplier Invitrogen™ Supplier No. PA542132

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: CDQLADRRSP VIEFWENKVL RLSGGLEVPG ALNWEVTLCL LACWVLVYFC Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

SLC6A8 (solute carrier family 6 member 8), also known as CT1 (creatine transporter 1), CRT or CRTR, is a 635 amino acid multi-pass plasma membrane protein that belongs to the sodium/neurotransporter (SNF) family. Expressed in a variety of tissues including kidney, skeletal muscle, heart, brain, prostate, testis and colon, SLC6A8 functions to transport creatine into and out of cells, specifically those found in brain and muscle tissue. Creatine, an organic acid, occurs naturally and is essential for supplying the energy needed for proper muscle and nerve function. Defects in the gene encoding SLC6A8 lead to cerebral creatine deficiency and are the cause of X-linked creatine deficiency syndrome, a rare disorder characterized by facial anomalies, seizures and mental retardation. Multiple isoforms of SLC6A8 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC6A8
Gene Accession No. P48029
Gene Alias AA589632; CCDS1; choline transporter; CHOT1; CHT1; Creat; creatine transporter; creatine transporter 1; creatine transporter SLC6A8 variant D; creatine transporter, solute carrier family 6, member 8; CRT; CRTR; CT1; CTR5; MGC87396; Na+ dependent transporter; SLC6A8; Sodium- and chloride-dependent creatine transporter 1; solute carrier family 6 (neurotransmitter transporter), member 8; solute carrier family 6 (neurotransmitter transporter, creatine), member 8; solute carrier family 6 member 8; solute carrier family 6, member 8
Gene Symbols SLC6A8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC6A8 (aa 200-249).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6535
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.