Learn More
Invitrogen™ SLC6A8 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598276
Product Code. 16541102
  • € 466.00 / 100µg
Description
SLC6A8 Polyclonal Antibody for ICC/IF, ELISA
SLC6A8 (solute carrier family 6 member 8), also known as CT1 (creatine transporter 1), CRT or CRTR, is a 635 amino acid multi-pass plasma membrane protein that belongs to the sodium/neurotransporter (SNF) family. Expressed in a variety of tissues including kidney, skeletal muscle, heart, brain, prostate, testis and colon, SLC6A8 functions to transport creatine into and out of cells, specifically those found in brain and muscle tissue. Creatine, an organic acid, occurs naturally and is essential for supplying the energy needed for proper muscle and nerve function. Defects in the gene encoding SLC6A8 lead to cerebral creatine deficiency and are the cause of X-linked creatine deficiency syndrome, a rare disorder characterized by facial anomalies, seizures and mental retardation. Multiple isoforms of SLC6A8 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC6A8
Polyclonal
Unconjugated
SLC6A8
AA589632; CCDS1; choline transporter; CHOT1; CHT1; Creat; creatine transporter; creatine transporter 1; creatine transporter SLC6A8 variant D; creatine transporter, solute carrier family 6, member 8; CRT; CRTR; CT1; CTR5; MGC87396; Na+ dependent transporter; SLC6A8; Sodium- and chloride-dependent creatine transporter 1; solute carrier family 6 (neurotransmitter transporter), member 8; solute carrier family 6 (neurotransmitter transporter, creatine), member 8; solute carrier family 6 member 8; solute carrier family 6, member 8
Rabbit
Protein G
RUO
6535
-20°C or -80°C if preferred
Liquid
ELISA, Immunocytochemistry
2.765 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
P48029
SLC6A8
Recombinant Human Sodium- and chloride-dependent creatine transporter 1 protein (582-635AA).