missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC7A11 Monoclonal Antibody (A7C6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544962
This item is not returnable.
View return policy
Description
SLC7A11 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
SLC7A11 (also known as XCT) is a member of a hetromeric, sodium-independent, anionic amino acid transport system. It is highly specific for cysteine and glutamate. SLC7A11 is a mediator of Kaposi sarcoma-associated herpes virus fusion and entry into cells. Increased expression of this protein in primary gliomas is associated with neuronal cell death.
Specifications
| SLC7A11 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9UPY5, Q9WTR6 | |
| SLC7A11 | |
| 293 cell line overexpressing SLC7A11. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| A7C6 | |
| Unconjugated | |
| SLC7A11 | |
| 9930009M05Rik; AI451155; Amino acid transport system xc-; calcium channel blocker resistance protein CCBR1; CCBR1; cysteine/glutamate transporter; cystine/glutamate transporter; SLC7A11; sodium independent anionic amino acid transport system; solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11; solute carrier family 7 (cationic amino acid transporter, y+ system), member 11; solute carrier family 7 member 11; solute carrier family 7, (cationic amino acid transporter, y+ system) member 11; sut; xCT | |
| Mouse | |
| Protein A | |
| RUO | |
| 23657, 26570 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction