missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93699-0.02ml
This item is not returnable.
View return policy
Description
SLC7A2 Polyclonal antibody specifically detects SLC7A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SLC7A2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin | |
| CAT2, CAT-2, cationic 2, cationic amino acid transporter, y+ system, low affinity cationic amino acid transporter 2, low-affinity cationic amino acid transporter-2, solute carrier family 7 (cationic amino acid transporter, y+ system), member 2, Solute carrier family 7 member 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 559-658 of human SLC7A2 (NP_001008539.3). PRNLSSPFIFHEKTSEF | |
| 0.02 mL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 6542 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction