Learn More
SLC7A5/LAT1 Antibody (EP3-1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20115MFV610
Product Code. 18545087
  • € 541.00 / 0.10mL
Description
SLC7A5/LAT1 Monoclonal antibody specifically detects SLC7A5/LAT1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC7A5/LAT1
Immunoprecipitation
Monoclonal
mFluor Violet 610 SE