missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93913-0.1ml
This item is not returnable.
View return policy
Description
SLC7A8 Polyclonal antibody specifically detects SLC7A8 in Human samples. It is validated for Western Blot
Specifications
| SLC7A8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| hLAT2, integral membrane protein E16H, large neutral amino acids transporter small subunit 2, LAT2L-type amino acid transporter 2, LPI-PC1, solute carrier family 7 (amino acid transporter, L-type), member 8, Solute carrier family 7 member 8, y+ system), member 8 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SLC7A8 (NP_036376.2). LFPTCFPPESGLRLLAAICLLLLTWVNCSSVRWATRVQDIFTAGKLLALALIIIMGIVQICKGEYFWLEPKNAFENFQEPDIGLVALAFLQGSFAYGGWNF | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 23428 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction