Learn More
SLC7A8 Mouse anti-Human, Clone: OTI7D6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500515
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SLC7A8 is sodium-independent, high-affinity transport of large neutral amino acids. It has higher affinity for L-phenylalanine than LAT1 but lower affinity for glutamine and serine. L-alanine is transported at physiological concentrations. SLC7A8 also plays a role in basolateral (re)absorption of neutral amino acids.Specifications
| SLC7A8 | |
| Monoclonal | |
| Unconjugated | |
| SLC7A8 | |
| LAT2, LPI-PC1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23428 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7D6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UHI5 | |
| SLC7A8 | |
| Full-length protein expressed in 293T cell transfected with human SLC7A8 expression vector | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.