missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC8B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101901
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC24A6.
SLC24A6 belongs to a family of potassium-dependent sodium/calcium exchangers that maintain cellular calcium homeostasis through the electrogenic counter transport of 4 sodium ions for 1 calcium ion and 1 potassium ion.
Specifications
| SLC8B1 | |
| Polyclonal | |
| Unconjugated | |
| SLC8B1 | |
| AF261233; Mitochondrial sodium/calcium exchanger protein; Na(+)/K(+)/Ca(2+)-exchange protein 6; Nckx6; NCLX; Slc24a6; Slc8b1; Sodium/calcium exchanger protein, mitochondrial; Sodium/potassium/calcium exchanger 6; sodium/potassium/calcium exchanger 6, mitochondrial; solute carrier family 24 (sodium/lithium/calcium exchanger), member 6; solute carrier family 24 (sodium/potassium/calcium exchanger), member 6; solute carrier family 24 member 6; solute carrier family 8 (sodium/lithium/calcium exchanger), member B1; solute carrier family 8 member B1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80024 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6J4K2 | |
| SLC8B1 | |
| A synthesized peptide derived from human SLC8B1(Accession Q6J4K2), corresponding to amino acid residues G493-V543. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction