missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC9A10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16610615 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16610615 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16610615 Supplier Invitrogen™ Supplier No. PA5104160

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC9A10.

SLC9A10 is a member of the sodium-hydrogen exchanger (NHE) family (see SLC9A1, MIM 107310) and is required for male fertility and sperm motility (Wang et al., 2003 [PubMed 14634667]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC9C1
Gene Accession No. Q4G0N8, Q6UJY2
Gene Alias Gm610; LOW QUALITY PROTEIN: sodium/hydrogen exchanger 10; Na(+)/H(+) exchanger 10; NHE; NHE-10; Slc9a10; SLC9C1; sNHE; Sodium/hydrogen exchanger 10; solute carrier family 10 member 10; Solute carrier family 9 member 10; Solute carrier family 9 member C1; solute carrier family 9, isoform 10; solute carrier family 9, member 10; solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1; spermNHE; sperm-NHE; Sperm-specific Na(+)/H(+) exchanger; sperm-specific sodium proton exchanger
Gene Symbols SLC9C1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC9C1(Accession Q4G0N8), corresponding to amino acid residues Q542-K592.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208169, 285335
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.