missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC9A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542501
This item is not returnable.
View return policy
Description
Peptide sequence: LGWGLRVAAA ASASSSGAAA EDSSAMEELA TEKEAEESHR QDSVSLLTFI Sequence homology: Dog: 100%; Goat: 85%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Zebrafish: 100%.
Na+/H+ exchangers (NHE) of mammalian cells are plasma membrane intrinsic proteins mediating exchange of N+ and H+ ions in various tissues. The NHE catalyzes the electroneural transport of extracellular Na+ for intracellular H+. They play a major role in regulation of intracellular pH (pHi) in addition to trans-cellular absorption of Na+, cell volume regulation and possibly in cell proliferation. These primary functions of the Na+/H+ exchanger have been related to many pathophysiological states, include hypertension, organ growth and hypertrophy, regression of cancer and renal intestinal disorders. At least 7 NHE isoforms (NHE1-7) have been cloned so far. They are all similar in their primary structure and predicted to have 10-12 transmembrane domains. The C-terminal domain of NHEs are predicted to be intracellular. NHE7 (human 725 aa, chromosome Xp11.4) is ubiquitously expressed, and predominantly localizes to the trans-golgi network. NHE7 mediates the influx of Na+ or K+ in exchange for H+. It is ~70% related to NHE6 but relatively less (~25%) homologous with other NHEs.
Specifications
| SLC9A7 | |
| Polyclonal | |
| Unconjugated | |
| SLC9A7 | |
| A530087D17Rik; hypothetical protein LOC554532; Na(+)/H(+) exchanger 7; Nhe7; NHE-7; nonselective sodium potassium/proton exchanger; SLC9A6; Slc9a7; Sodium/hydrogen exchanger 7; solute carrier family 9 (sodium/hydrogen exchanger), isoform 7; solute carrier family 9 (sodium/hydrogen exchanger), member 7; solute carrier family 9 member 7; solute carrier family 9 member A7; solute carrier family 9, subfamily A (NHE7, cation proton antiporter 7), member 7; wu:fk03b09; zgc:101088; zgc:113878 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84679 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96T83 | |
| SLC9A7 | |
| Synthetic peptide directed towards the N-terminal of human SLC9A7 (aa 29-78). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction