missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC9A9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101895
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC9A9.
This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.
Specifications
| SLC9A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC9A9 | |
| 5730527A11Rik; 9930105B05; AI854429; AUTS16; FLJ35613; Na(+)/H(+) exchanger 9; Nbla00118; NHE9; NHE-9; putative protein product of Nbla00118; RGD1560736; Slc9a9; Sodium/hydrogen exchanger 9; sodium/proton exchanger NHE9; solute carrier family 9 (sodium/hydrogen exchanger), isoform 9; solute carrier family 9 (sodium/hydrogen exchanger), member 9; solute carrier family 9 member 9; solute carrier family 9 member A9; solute carrier family 9, subfamily A (NHE9, cation proton antiporter 9), member 9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 285195, 331004 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BZ00, Q8IVB4 | |
| SLC9A9 | |
| A synthesized peptide derived from human SLC9A9(Accession Q8IVB4), corresponding to amino acid residues I172-I222. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction