missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLCO2A1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA553083

Product Code. 15731031

  • € 596.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PSTSSSIHPQ SPACRRDCSC PDSIFHPVCG DNGIEYLSPC HAGCSNINMS SATSKQLIYL NCSCVTGGSA SAKTGSCPVP CAH Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

Prostaglandin transporter (PGT), also known as SLCO2A1, has been identified as a functional prostaglandin (PG) carrier that mediates either the release of newly-synthesized PGs by cells and/or the uptake of PGs prior to intracellular oxidation. Northern blot analysis revealed that PGT mRNA has broad tissue distribution, suggesting its role in the transport and/ or metabolic clearance of PG in diverse tissues (8787677).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLCO2A1
Polyclonal
Unconjugated
SLCO2A1
2310021C19Rik; MATR1; matrin F/G 1; mPgt; OATP2A1; PGT; PHOAR2; Prostaglandin transporter; Slc21a2; Slco2a1; solute carrier family 21 (prostaglandin transporter), member 2; solute carrier family 21 member 2; Solute carrier organic anion transporter family member 2A1; solute carrier organic anion transporter family, member 2A1
Rabbit
Antigen affinity chromatography
RUO
6578
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q92959
SLCO2A1
Recombinant protein corresponding to Human SLCO2A1. Recombinant protein control fragment (Product #RP-90771).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.