Learn More
SLFN14 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82010PCP
This item is not returnable.
View return policy
Description
SLFN14 Polyclonal antibody specifically detects SLFN14 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| SLFN14 | |
| schlafen family member 14 | |
| Immunology |
| Western Blot | |
| RUO | |
| Purified |