missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLFNL1 Antibody (OTI1G2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74223H
This item is not returnable.
View return policy
Description
SLFNL1 Monoclonal specifically detects SLFNL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SLFNL1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| SLFNL1 | |
| Full length recombinant protein of human SLFNL1 (NP_659427) produced in HEK293T cells. | |
| 0.1 mL | |
| 200172.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1G2 | |
| Western Blot, Immunohistochemistry | |
| FLJ23878, MGC43873, schlafen-like 1, schlafen-like protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction