Learn More
ibidi μ-Slide 1 Well ibiTreat
Description
- No. 1.5 polymer coverslip
- Tissue culture-treated
- Sterilized
- Individually packed
Specifications
Specifications
| Type | Chamber Slide |
| Surface Treatment | ibiTreat |
| Material | Polymer |
| For Use With (Application) | Cell Culture, High-end Microscopy |
| No. per Case | 15/Cs. |
| No. per Pack | 1 Ea. |
| Sterility | Sterile |