Learn More
ibidi μ-Slide 1 Well Untreated
Description
- No. 1.5 polymer coverslip
- Hydrophobic
- Sterilized
- Individually packed
Specifications
Specifications
| Type | Chamber Slide |
| Surface Treatment | Untreated |
| Material | Polymer |
| For Use With (Application) | Cell Culture, High-end Microscopy |
| No. per Case | 120/Cs. |
| No. per Pack | 8/Tray, 15 Trays |
| Sterility | Sterile |