Learn More
Ibidi μ-Slide 15 Well 3D Glass Bottom
Click to view available options
:
μ-Slide
Description
- A chambered coverslip with 15 wells for 3D cell culture, tube formation assays, and immunofluorescence staining
- bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
- sterilized
Specifications
Specifications
| Type | Coverglass |
| Sterility | Sterile |