Learn More
Ibidi μ-Slide 15 Well 3D Glass Bottom
Product Code. 16863993
Encompass_Preferred
Click to view available options
:
μ-Slide
Product Code. 16863993
Brand: Ibidi 81507
Only null left
Add to basket
Add to basket
μ-Slide 15 Well 3D Glass Bottom is a chambered coverslip with 15 wells.
  • A chambered coverslip with 15 wells for 3D cell culture, tube formation assays, and immunofluorescence staining
  • bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
  • sterilized
Specifications
Type Coverglass
Sterility Sterile