missing translation for 'onlineSavingsMsg'
Learn More

ibidi μ-Slide 18 Well ibiTreat

Product Code. 30130499
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
90/Cs.
Unit Size:
Pack of 120
Pack of 90
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case Unit Size
30130499 120/Cs. Pack of 120
30130514 90/Cs. Pack of 90
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30130499 Supplier ibidi Supplier No. 81816120
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A chambered coverslip with 18 wells.

  • A chambered coverslip for cell culture, immunofluorescence, and high-end microscopy
  • Bottom material: No. 1.5 Polymer Coverslip
  • Tissue culture treated
  • Sterilized

Specifications

Type Chamber Slide
Wells 18
Surface Treatment ibiTreat
Material Polymer
For Use With (Application) Cell Culture
No. per Case 120/Cs.
No. per Pack 8/Tray, 15 Trays
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.