Learn More
Ibidi μ-Slide 8 Well High ibiTreat, Bulk Pack
Product Code. 17236203
Encompass_Preferred
Click to view available options
:
μ-Slide
Product Code. 17236203
Brand: Ibidi 8080690
Only null left
Add to basket
Add to basket
μ-Slide 8 Well High ibiTreat, Bulk Pack is a chambered coverslip with 8 individual wells and high walls.
  • A chambered coverslip with 8 individual wells and high walls for cell culture, immunofluorescence, and high-end microscopy
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • individually packed
  • sterilized
Specifications
Type Chambered Coverglass
Wells 8