Learn More
Ibidi μ-Slide 8 Well high μ-Pattern RGD, sqr30, pit110, hex
Click to view available options
:
μ-Slide
Description
- A chambered coverslip with 8 wells and micropatterned surface
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
- sterilized
Specifications
Specifications
| Type | Microscope Slide |
| Well Count | 8 |
| Quantity | 10 |