Learn More
Ibidi μ-Slide 8 Well high μ-Pattern RGD, sqr30, pit110, hex
Product Code. 17894784
Encompass_Preferred
Click to view available options
:
μ-Slide
Product Code. 17894784
Brand: Ibidi 83803
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
μ-Slide 8 Well high μ-Pattern RGD, sqr30, pit110, hex is an 8 well chambered coverslip with micropatterned surface.
  • A chambered coverslip with 8 wells and micropatterned surface
  • bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
  • sterilized
Specifications
Type Microscope Slide
Well Count 8
Quantity 10