Learn More
Epredia™ Slide Baskets and Hangers for Shandon™ ClearVue™ Coverslippers, Hangers for Sakura DRS 2000-compatible ClearVue baskets
Shop All Epredia ProductsDescription
- Allow equipment compatibility with a variety of different stainers
- Can be loaded in mix-or-match fashion
- Sold in sets of five
Specifications
Specifications
| For Use With (Equipment) | Shandon ClearVue Coverslipper |
| Description | Hangers for Sakura DRS 2000-Compatible ClearVue Baskets |