Learn More
Ibidi μ-Slide III 3in1 ibiTreat
Product Code. 17296163
Encompass_Preferred
Click to view available options
:
μ-Slide
Product Code. 17296163
Brand: Ibidi 80316
Only null left
Add to basket
Add to basket
μ-Slide III 3in1 ibiTreat is a μ-Slide for switchable chemotactic gradients.
  • A channel slide for switchable chemotactic gradients that merges 3 separated liquids into one channel
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Type Microscope Slide
Quantity 15