Learn More
ibidi μ-Slide Tissue Engineering Untreated
Product Code. 31941062
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
15/Cs.
90/Cs.
No. per Pack:
1 Ea.
8/Tray, 15 Trays
Unit Size:
Pack of 120
Pack of 15
Pack of 90
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case No. per Pack Unit Size
31941062 15/Cs. 1 Ea. Pack of 15
31940922 90/Cs. 1 Ea. Pack of 90
31941027 120/Cs. 8/Tray, 15 Trays Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31941062 Supplier ibidi Supplier No. 80231
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Microfluidic channel slide for tissue model and organ-on-a-chip applications
  • A μ-Slide for cell culture and high-end microscopy
  • No. 1.5 polymer coverslip
  • Hydrophobic
  • Sterilized
Specifications
Type Chamber Slide
Surface Treatment Untreated
Material Polymer
For Use With (Application) Cell Culture, High-end Microscopy
No. per Case 15/Cs.
No. per Pack 1 Ea.
Sterility Sterile