Learn More
ibidi μ-Slide VI 0.4 Bioinert
Description
- Superior to the standard ultra-low attachment (ULA) surfaces - stable, biologically inert surface for long-term experiments without any cell or biomolecule adhesion
- No. 1.5 polymer coverslip
- Surface passivation with Bioinert
- Sterilized
Specifications
Specifications
| Type | Chamber Slide |
| Surface Treatment | Bioinert |
| Material | Polymer |
| No. per Case | 90/Cs. |
| No. per Pack | 1 Ea. |
| Sterility | Sterile |