Learn More
ibidi μ-Slide Y-shaped ibiTreat
Description
- A channel slide with bifurcations of 30 and 45 degree for bifurcation studies and blood vessel simulation
- Easy to connect with tubing and pump systems via female Luers
- Bottom material: No. 1.5 Polymer Coverslip
- Tissue culture treated
- Sterilized
Specifications
Specifications
| Type | Chamber Slide |
| Surface Treatment | ibiTreat |
| Material | Polymer |
| No. per Case | 90/Cs. |
| No. per Pack | 1 Ea. |
| Sterility | Sterile |