missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLIRP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146148
This item is not returnable.
View return policy
Description
Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: antigen standard for C14orf156 (transient overexpression lysate), human liver protein.
SLIRP (SRA stem-loop interacting RNA binding protein) is expressed in normal and tumor tissues, but the majority of the endogenous SLIRP resides in the mitochondria. SLIRP may regulate mitochondrial functions and has the potential to shut down estrogen in breast cancer cells and testosterone in prostate cancer cells. SLIRP has also been shown to turn down genes involved in energy metabolism.
Specifications
| SLIRP | |
| Polyclonal | |
| Unconjugated | |
| SLIRP | |
| 1810035L17Rik; C14orf156; DC23; DC50; PD04872; SLIRP; SRA stem-loop interacting RNA binding protein; SRA stem-loop-interacting RNA-binding protein, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 380773, 81892 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9D8T7, Q9GZT3 | |
| SLIRP | |
| A synthetic peptide made to a C-terminal region of human SLIRP protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction