Learn More
Invitrogen™ SLIT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595763
Description
Immunogen sequence: RNLYINNELQ DFTKTQMKPG VVPGCEPCRK LYCLHGICQP NATPGPMCHC EAGWVGLHCD QPADGPCHGH KCVHGQCVPL DALSYSCQCQ DGYSGALCNQ AGALAEPCRG LQCLHGHCQA SGTKGAHCVC DPGFSGELCE QESECRGDPV RDFHQVQRGY AICQTTRPLS WVECRGSCPG QGCCQGLRLK RRKFTFECSD GTSFAEEVEK PTKCGCALCA; Positive Samples: 293T.
SLIT1 is thought to act as molecular guidance cue in cellular migration, and function appears to be mediated by interaction with roundabout homolog receptors.
Specifications
| SLIT1 | |
| Polyclonal | |
| Unconjugated | |
| SLIT1 | |
| KIAA0813; Megf4; mKIAA0813; Multiple EGF-like domains protein 4; multiple epidermal growth factor-like domains 4; multiple epidermal growth factor-like domains protein 4; SLIL1; slit guidance ligand 1; slit homolog 1; slit homolog 1 (Drosophila); slit homolog 1 protein; SLIT1; Slit-1; SLIT3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6585 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.43 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75093 | |
| SLIT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1315-1534 of human SLIT1 (NP_0030522). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.