missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLIT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20217-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to SLIT2.
Slit homolog 2 proteins (SLIT2) are ligands for the Roundabout (Robo) receptors and act as molecule guidance cues in cell migration. Slit1 and Slit2 are essential for midline guidance in the forebrain by acting as repulsive signals preventing inappropriate midline crossing by axons projecting from the olfactory bulb. Slit2 protein is expressed in fetal lung and kidney, and adult spinal cord. Weak expression is also noticed in adult adrenal gland, thyroid, and trachea. SLIT2 is a target for repression by EZH2 and is up-regulated in prostate cancer.Specifications
| SLIT2 | |
| Polyclonal | |
| Unconjugated | |
| SLIT2 | |
| SLIL3, Slit 2, slit homolog 2 (Drosophila), Slit homolog 2 protein, SLIT2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20563, 360272, 9353 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O94813, Q9R1B9, Q9WVC1 | |
| Slit2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu