missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK1 Antibody (1), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89435DL594
This item is not returnable.
View return policy
Description
SLITRK1 Monoclonal antibody specifically detects SLITRK1 in Human samples. It is validated for ELISA
Specifications
| SLITRK1 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA | |
| FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SLITRK1 (NP_443142.1; Met 1-Ser 616). | |
| 0.1 mL | |
| Neuroscience | |
| 114798 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction