Learn More
Invitrogen™ SLITRK2 Monoclonal Antibody (312420)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561193
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This intronless gene is expressed in the hippocampus and maps close to a candidate region for several X-linked mental retardation (XLMR) syndromes. It is conserved in primates, cow, and horse, but not found in mouse and rat. The exact function of this gene is not known, but on the basis of its physical location and expression pattern, it is proposed to have an important function in the brain. [provided by RefSeq, Jan 2010].
Specifications
| SLITRK2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9H156 | |
| SLITRK2 | |
| Mouse myeloma cell line NS0-derived recombinant human SLITRK2, Thr19-Pro617. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 312420 | |
| Unconjugated | |
| SLITRK2 | |
| A930040J07; CXorf2; KIAA1854; RGD1561361; SLIT and NTRK like family member 2; SLIT and NTRK-like family, member 2; SLIT and NTRK-like protein 2; slit and trk like gene 2; SLITL1; slit-like 1; Slitrk2; UNQ9197/PRO34756 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 84631 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |