missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76875-0.025mg
This item is not returnable.
View return policy
Description
SLITRK4 Polyclonal specifically detects SLITRK4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SLITRK4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DKFZp547M2010, SLIT and NTRK-like family, member 4, SLIT and NTRK-like protein 4, slit and trk like gene 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 139065 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Q8IW52 | |
| SLITRK4 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human Slitrk4. The immunogen is located within amino acids 30 - 80 of Slitrk4. Amino Acid Squence: SVYRPNQLKPPWSN | |
| 0.025 mg | |
| Primary | |
| This antibody is predicted to have no cross-reactivity to other Slitrk proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction