missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21150-1-AP-20UL
This item is not returnable.
View return policy
Description
SLIT and NTRK-like family 6 (Slitrk6) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Slitrk6 expression has been observed in tissues such as tongue, lung, gastrointestinal tract, and pancreas. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk6 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth. At least two isoforms of Slitrk6 are known to exisit.Specifications
| SLITRK6 | |
| Polyclonal | |
| Unconjugated | |
| SLITRK6 | |
| SLIT and NTRK like protein 6, SLITRK6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 239250, 290467, 84189 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8C110, Q9H5Y7 | |
| Slitrk6 | |
| SLITRK6 Fusion Protein Ag15701 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction