missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLK Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA553380

Product Code. 15741321

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DLVSQETGEK EANIQAVDSE VGLTKEDTQE KLGEDDKTQK DVISNTSDVI GTCEAADVAQ KVDEDSAEDT QSNDGKEVVE VGQKLINKPM VGPEAGGTKE VPIKEIVEMN EIEEGKNKEQ AINSSENIMD INEEPGTTEG EE Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 48%.

SLK acts as a novel effector of cytoskeletal remodeling and apoptosis, and seems to be ubiquitously expressed. SLK mediates apoptosis and actin stress fiber dissolution.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLK
Polyclonal
Unconjugated
SLK
9A2; AV021402; AW411554; bA16H23.1; CTCL tumor antigen se20-9; Etk4; hSLK; Kiaa0204; Long Ste20-like Kinase; LOSK; mKIAA0204; mSLK; se20-9; serine/threonine kinase 2; serine/threonine-protein kinase 2; Sk2; Slk; SMAK; SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrose nonfermenting like kinase; SNF1 sucrose nonfermenting like kinase; STE20 like kinase; STE20-like kinase; STE20-like serine/threonine-protein kinase; STE20-related kinase; STE20-related kinase SMAK; Ste20-related serine/threonine kinase; STE20-related serine/threonine-protein kinase; Stk2
Rabbit
Antigen affinity chromatography
RUO
9748
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H2G2
SLK
Recombinant protein corresponding to Human SLK. Recombinant protein control fragment (Product #RP-91422).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.