missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLMAP Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06131MFV500
This item is not returnable.
View return policy
Description
SLMAP Polyclonal antibody specifically detects SLMAP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLMAP | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| FLJ42206, KIAA1601Sarcolemmal-associated protein, MGC138760, sarcolemma associated protein, sarcolemmal membrane-associated protein, SLAPMGC138761 | |
| Produced in rabbits immunized with E. coli-derived Human SLMAP fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7871 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction