Learn More
Witeg™ Sloping Recovery Head with Thermometer Socket
Product Code. 10164904
Change view
Click to view available options
Cone Size:
Flask ST 19/26 Condenser ST 19/26
Flask ST 29/32 Condenser ST 19/26
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size unitSize
10164904 Flask ST 29/32 Condenser ST 19/26 Each
10536384 Flask ST 19/26 Condenser ST 19/26 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10164904 Supplier Witeg™ Supplier No. 1 010 006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replace broken or worn recovery heads with these quality replacement parts. Witeg™ Sloping Recovery Head with Thermometer Socket is designed with a flask cone, a condenser cone, and a thermometer socket for easy temperature measurements.
  • Sloping arm.
  • Thermometer socket.
  • Flask cone.
  • Condenser cone.
Specifications
Item Description With thermometer socket
Type Sloping
Cone Size Flask ST 29/32 Condenser ST 19/26
Standard Taper Joint No Thermometer Socket ST 14/23