Learn More
Witeg™ Sloping Recovery Head with Thermometer Socket
Description
- Sloping arm.
- Thermometer socket.
- Flask cone.
- Condenser cone.
Specifications
Specifications
| Item Description | With thermometer socket |
| Type | Sloping |
| Cone Size | Flask ST 29/32 Condenser ST 19/26 |
| Standard Taper Joint No | Thermometer Socket ST 14/23 |