Learn More
Hoefer™ Slotted Gaskets for SE 400 and SE 600 Series Units
Click to view available options
Quantity:
1/Pk.
2/Pk.
Unit Size:
Each
Pack of 2
Description
- For use with SE 400 and SE 600 Series Units
Specifications
Specifications
| Color | Gray |
| For Use With (Equipment) | SE 400, SE 600 Series Vertical Electrophoresis Unit |
| Type | Slotted Gasket |
| Material | Silicone Rubber |
| Quantity | 2/Pk. |