Learn More
Slow Skeletal Muscle Myosin Heavy Chain Antibody (M14), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50297JF646
Product Code. 18115288
  • € 437.00 / 0.10mL
Description
Slow Skeletal Muscle Myosin Heavy Chain Monoclonal specifically detects Slow Skeletal Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Slow Skeletal Muscle Myosin Heavy Chain
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
purified human skeletal muscle myosin
0.1 mL
Primary
Chicken, Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
M14
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Mouse
Protein A purified
Cardiovascular Biology, Cellular Signaling, Developmental Biology, Stem Cells
Specific for slow skeletal muscle myosin heavy chain (SM1).
Store at 4°C in the dark.