Learn More
SLP-2 Antibody (5D10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43042
This item is not returnable.
View return policy
Description
SLP-2 Monoclonal antibody specifically detects SLP-2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| SLP-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 5D10 | |
| Western Blot 1:2000 - 1:10000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunoprecipitation 1:500 - 1:1000 | |
| EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2 | |
| stomatin(EPB72)-like 2 - (Uniprot# Q9UJZ1) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 30968 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |