missing translation for 'onlineSavingsMsg'
Learn More

SLP-76/LCP2 Antibody, Novus Biologicals™

Product Code. p-200049497 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18450881 25 μL 25µL
18424041 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18450881 Supplier Novus Biologicals Supplier No. NBP18703225ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLP-76/LCP2 Polyclonal antibody specifically detects SLP-76/LCP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLP-76/LCP2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias lymphocyte cytosolic protein 2, lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of76kDa), lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of76kD), SH2 domain-containing leukocyte protein of 76 kDa, SH2 domain-containing leukocyte protein of 76kD, SLP-76, SLP-76 tyrosine phosphoprotein, SLP7676 kDa tyrosine phosphoprotein
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: PNEEEEAPVEDDADYEPPPSNDEEALQNSILPAKPFPNSNSMYIDRPPSGKTPQQPPVPPQRPMAALPPPPAGRNHSPLPPPQTNHEEPSRSRNHKT
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3937
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.