missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLPI Antibody (001), DyLight 755 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89438IR
This item is not returnable.
View return policy
Description
SLPI Monoclonal antibody specifically detects SLPI in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
| SLPI | |
| Monoclonal | |
| DyLight 755 | |
| ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SLPI (P03973; Met1-Ala132). | |
| 0.1 mL | |
| Cancer | |
| 6590 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunofluorescence | |
| 001 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction