missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLU7 Monoclonal Antibody (OTI7B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810916S
This item is not returnable.
View return policy
Description
SLU7 Monoclonal antibody specifically detects SLU7 in Human samples. It is validated for Western BlotSpecifications
| SLU7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95391 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10569 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI7B7 | |
| Unconjugated | |
| SLU7 | |
| 9G8; AU018913; D11Ertd730e; D3Bwg0878e; hSlu7; MGC9280; Pre-mRNA-splicing factor SLU7; SLU7; SLU7 homolog, splicing factor; SLU7 splicing factor homolog; SLU7 splicing factor homolog (S. cerevisiae); splicing factor; step II splicing factor SLU7; zinc knuckle motif containing | |
| Human recombinant protein fragment corresponding to amino acids 1-300 of human SLU7 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction