Learn More
Invitrogen™ SLUG Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5120095
Product Code. 17761494
  • € 514.00 / 100µL
Description
Positive test controls include: Mouse spleen, Mouse heart. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MPRSFLVKKH FNASKKPNYS ELDTHTVIIS PYLYESYSMP VIPQPEILSS GAYSPITVWT TAAPFHAQLP NGLSPLSGYS SSLGRVSPPP PSDTSSKDHS GSESPISDEE ERLQSKLSDP HAIEAEKFQC NLCNKTYSTF SGLAKHKQLH.
Slug, a member of the Snail family of C2H2-type zinc finger transcription factors, was initially identified to be involved in epithelial-mesenchymal transitions as well as the formation of the neural tube during vertebrate embryogenesis. Like Snail, Slug transcription can be induced by growth factors such as FGF, BMP, and TGF-beta. Once expressed, Slug will bind E-box regions of promoters and repress transcription of genes such as E-cadherin and Claudin-1. More recently, its expression in breast, esophogeal, and colorectal carcinomas has been correlated with poor prognosis for survival. Furthermore, Slug can protect hemopoietic progenitor cells from radiation-induced apoptosis by repressing the p53-mediated transcription of Puma, a BH3-only antagonist of the anti-apoptotic members of the Bcl-2 family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLUG
Polyclonal
Unconjugated
SNAI2
MGC10182; neural crest transcription factor Slug; Protein snail homolog 2; SLUG; slug (chicken homolog), zinc finger protein; Slug, chicken homolog; Slugh; SLUGH1; Snai2; snail family transcriptional repressor 2; snail family zinc finger 2; snail homolog 2; Snail2; snail-like protein 2; WS2D; Zinc finger protein SNAI2; zinc finger transcription factor snail 2
Rabbit
Affinity Chromatography
RUO
20583, 6591
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.02 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43623, P97469
SNAI2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human Slug (NP_0030591).
100 μL
Primary
Human, Mouse