missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLX1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546622
This item is not returnable.
View return policy
Description
Peptide sequence: QEEPGQLLPL EGQCPCCEKS LLWGDLIWLC QMDTEKEVED SELEEAHWTD Sequence homology: Horse: 90%; Human: 100%; Yeast: 79%.
SLX1 encodes a protein that is an important regulator of genome stability. The protein represents the catalytic subunit of the SLX1-SLX4 structure-specific endonuclease, which can resolve DNA secondary structures that are formed during repair and recombination processes. Two identical copies of this gene are located on the p arm of chromosome 16 due to a segmental duplication. SLZ1A represents the more centromeric copy and SLZ1B represents the more telomeric copy.
Specifications
| SLX1B | |
| Polyclonal | |
| Unconjugated | |
| SLX1A | |
| SLX1A; SLX1B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 548593, 79008 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BQ83 | |
| SLX1A, SLX1B | |
| Synthetic peptide directed towards the C-terminal of human SLX1B (aa 222-271). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction