missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLX4 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28680JF646
This item is not returnable.
View return policy
Description
SLX4 Polyclonal antibody specifically detects SLX4 in Human samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)
Specifications
| SLX4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84464 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Janelia Fluor 646 | |
| BTB (POZ) domain containing 12, BTB/POZ domain-containing protein 12, BTBD12structure-specific endonuclease subunit SLX4, KIAA1784MUS312, KIAA1987FANCP, SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae) | |
| The immunogen recognized by this antibody maps to a region between residue 1784 to 1834 of human SLX4 using the numbering given in entry NP_115820.2 (GeneID 84464). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction