missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLX4 Interacting Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30208
This item is not returnable.
View return policy
Description
SLX4 Interacting Protein Polyclonal antibody specifically detects SLX4 Interacting Protein in Human samples. It is validated for Immunoprecipitation
Specifications
| SLX4 Interacting Protein | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 128710 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| bA204H22.1, bA254M13.1, C20orf94 chromosome 20 open reading frame 94, chromosome 20 open reading frame 94, dJ1099D15.3 | |
| Between 175 and 225 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction