missing translation for 'onlineSavingsMsg'
Learn More

SM-164 Hydrochloride, MedChemExpress

Product Code. 16499497
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499497

Brand: Medchem Express HY15989A5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SM-164 Hydrochloride is a cell-permeable Smac mimetic compound. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains with an IC50 value of 1.39 nM and functions as an extremely potent antagonist of XIAP.

SM-164 Hydrochloride is a cell-permeable Smac mimetic compound. SM-164 binds to XIAP protein containing both the BIR2 and BIR3 domains with an IC50 value of 1.39 nM and functions as an extremely potent antagonist of XIAP.

Specifications

Chemical Name or Material SM-164 Hydrochloride
Color Off-White
Purity Grade Notes Research
Molecular Formula C62H85ClN14O6
Quantity 5 mg
Solubility Information H2O : ≥ 106 mg/mL (91.55 mM)
SMILES C[C@H](NC)C(N[C@H]1CCCC[C@](CC[C@H]2C(N[C@@H](C3=CC=CC=C3)C4=CN(CCCCC5=CC=C(CCCCN6N=NC([C@@H](NC([C@@H]7CC[C@@](CCCC[C@@H]8NC([C@@H](NC)C)=O)([H])N7C8=O)=O)C9=CC=CC=C9)=C6)C=C5)N=N4)=O)([H])N2C1=O)=O.Cl
Molecular Weight (g/mol) 1157.88
Formula Weight 1157.88
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.