missing translation for 'onlineSavingsMsg'
Learn More

SMAC/Diablo Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 167.00 - € 507.00

Specifications

Antigen SMAC/Diablo
Concentration 1 mg/mL
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/ Immunofluorescence 10 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18518802
View Documents
Novus Biologicals
NBP1-76772-0.025mg
0.025 mg
€ 167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18506660
View Documents
Novus Biologicals
NBP1-76772
0.1 mg
€ 507.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SMAC/Diablo Polyclonal antibody specifically detects SMAC/Diablo in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

SMAC/Diablo
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/ Immunofluorescence 10 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse, Rat
AF203914
56616
IgG
Ion exchange chromatography
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Apoptosis, Cancer, Cell Biology, Cellular Markers, Mitochondrial Markers, Tumor Suppressors
PBS
0610041G12Rik, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial
Antibody was raised against a 16 amino acid peptide near the carboxy terminus of murine Smac. The immunogen is located within the last 50 amino acids of Smac. Amino Acid Squence: SDEGADQEEEAYLRED
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.