missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD1/5/9 Antibody (RM487), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-18834
This item is not returnable.
View return policy
Description
SMAD1/5/9 Monoclonal antibody specifically detects SMAD1/5/9 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SMAD1/5/9 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| RM487 | |
| Western Blot 1:1000-1:2000 | |
| BSP1, BSP-1, hSMAD1, JV4-1SMAD, mothers against DPP homolog 1 (Drosophila), MAD homolog 1, MADH1JV41, MADR1MAD, mothers against decapentaplegic homolog 1 (Drosophila), Mad-related protein 1, mothers against decapentaplegic homolog 1, Mothers against DPP homolog 1, SMAD family member 1SMAD 1, SMAD, mothers against DPP homolog 1, Smad1, TGF-beta signaling protein 1, transforming growth factor-beta signaling protein 1 | |
| A phospho-peptide corresponding to human Phospho-SMAD1 [Ser463/465]/ SMAD5 [Ser463/465]/ SMAD9 [Ser465/467] | |
| 100 μg | |
| Cancer, Diabetes Research, Stem Cell Signaling Pathway | |
| 4086 | |
| Store at -20°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction