missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD1 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507244S
This item is not returnable.
View return policy
Description
SMAD1 Monoclonal antibody specifically detects SMAD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SMAD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15797 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4086 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C5 | |
| Unconjugated | |
| SMAD1 | |
| AI528653; BSP1; BSP-1; dwarfin-A; Dwf-A; DWFC; hSMAD1; JV41; JV4-1; JV5-1; MAD (mothers against decapentaplegic, Drosophila) homolog 1; MAD homolog 1; MAD homolog1 (mothers against decapentaplegic, Drosophila); MAD, mothers against decapentaplegic homolog 1; Madh1; MADH5; MADR1; Mad-related protein 1; Mlp1; mMad1; mothers against decapentaplegic homolog 1; mothers against DPP homolog 1; mothers-against-DPP-related 1; Mothers-against-DPP-related-1; MusMLP; OTTHUMP00000220456; SMAD; Smad 1; SMAD family member 1; SMAD, mothers against DPP homolog 1; Smad1; SMAD5; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1 | |
| Full length human recombinant protein of human SMAD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction